Dichloroacetic acid (DCA) ≥99%, Technical Grade

Supplier: Thermo Scientific Chemicals

Synonyms: DCA Deblock, DCA

473140010 473140025
TS473140010EA 130 USD
TS473140010 TS473140025
Dichloroacetic acid (DCA) ≥99%, Technical Grade
Dichloroacetic acid (DCA)
Formula: C₂H₂Cl₂O₂
MW: 128.94 g/mol
Boiling Pt: 194
Melting Pt: -4
MDL Number: MFCD00004223
CAS Number: 79-43-6
UN: 1764
ADR: 8 II
Merck Index: 13,03075

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR