Deuterium chloride 20% (w/w) in D₂O 99.5 atom % D for NMR spectroscopy

Supplier: Thermo Scientific Chemicals

464140250
TS464140250EA 93.1 USD
TS464140250
Deuterium chloride 20% (w/w) in D₂O 99.5 atom % D for NMR spectroscopy
Deuterium chloride
Formula: DCl
MW: 37.45 g/mol
MDL Number: MFCD00044642
CAS Number: 7698-05-7
UN: 1789

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR