Dimethyl sulfoxide-[D₆] 99.5 atom % D for NMR spectroscopy

Supplier: Thermo Scientific Chemicals

Synonyms: Dimethyl sulfoxide-[²H₆]Hexadeuterodimethyl sulfoxideDMSO-D6DMSO-d6Dimethyl sulphoxide-d6Dimethyl sulfoxide-d6Dimethyl sulphoxide-[²H₆]Hexadeuterodimethyl sulphoxide

464071000
TS464071000EA 303 USD
TS464071000
Dimethyl sulfoxide-[D₆] 99.5 atom % D for NMR spectroscopy
Dimethyl sulphoxide-[D6]
Formula: D₃CS(O)CD₃
MW: 84.09 g/mol
MDL Number: MFCD00002090
CAS Number: 2206-27-1
UN: 1993

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR