Hydrobromic acid 33% (w/w) in glacial acetic acid, pure

Supplier: Thermo Scientific Chemicals
Danger

123180010 123180025
TS123180010EA 213 USD
TS123180010 TS123180025
Hydrobromic acid 33% (w/w) in glacial acetic acid, pure
Hydrogen bromide
Formula: HBr
Density: 1.42 g/cm³
Flash Pt: 39 °C
Storage Temperature: Ambient
MDL Number: MFCD00011323
CAS Number: 10035-10-6
EINECS: 233-113-0
UN: 2920
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear yellow to yellow-orange Solution (May darken on storage)
Water ≤0.5 % (K.F.)
Titration with NaOH 31.5 to 36.5 % (HBr)
Particle size ≥50 % (> 32 µm)
Residue after ignition ≤0.05 %
pH 5 to 7
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR