Dimethyl sulfoxide-[D₆] 100.0 atom % D, > 99.96 atom % D for NMR spectroscopy
Supplier: Thermo Scientific Chemicals
Synonyms:
Dimethyl sulfoxide-[²H₆]
, Hexadeuterodimethyl sulfoxide
, DMSO-D6
, DMSO-d6
, Dimethyl sulphoxide-d6
, Dimethyl sulfoxide-d6
, Dimethyl sulphoxide-[²H₆]
, Hexadeuterodimethyl sulphoxide
169020250
TS169020250EA
216
USD
TS169020250
Dimethyl sulfoxide-[D₆] 100.0 atom % D, > 99.96 atom % D for NMR spectroscopy
Dimethyl sulphoxide-[D6]
Formula:
D₃CS(O)CD₃ MW: 84.09 g/mol Boiling Pt: 189 °C (5 mmHg) Melting Pt: 20.2 °C Density: 1.19 g/cm³ Flash Pt: 88 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00002090 CAS Number: 2206-27-1 EINECS: 218-617-0 |
Specification Test Results
Appearance | Clear colorless Liquid |
Water | ≤0.02 % (Coulometric) |
D-Enrichment | ≥99.95 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...