Orthoboric acid 99+%, extra pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
180570010
180570025
180570100
TS180570010EA
70
USD
TS180570010
TS180570025
TS180570100
Orthoboric acid 99+%, extra pure
ortho-Boric acid
Formula:
B(OH)₃ Melting Pt: 169 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00011337 CAS Number: 10043-35-3 EINECS: 233-139-2 REACH: 01-2119486683-25 |
Specification Test Results
Appearance | White Powder or crystals or granules |
Titration with NaOH | ≥99.0 % |
Chloride (Cl) | ≤10 ppm |
Sulfate (SO4) | ≤145 ppm |
Iron (Fe) | ≤10 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...