1,2-Dichlorobenzene 99%, pure

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: o-Dichlorobenzene

113180010 113180025
TS113180010EA 115 USD
TS113180010 TS113180025
1,2-Dichlorobenzene 99%, pure
1,2-Dichlorobenzene
Formula: C₆H₄Cl₂
MW: 147 g/mol
Boiling Pt: 180.5
Melting Pt: -17
Density: 1.3 g/cm³
Flash Pt: 67 °C
Storage Temperature: Ambient
MDL Number: MFCD00000535
CAS Number: 95-50-1
EINECS: 202-425-9
UN: 1591
ADR: 6.1 III
Merck Index: 13,03081

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5500 to 1.5520 (20°C, 589 nm)
GC ≥98.5 %
Suitability for use as a laser dye
Molar extinction coefficient ≥105000 (lambda max)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR