Tetrachloroethylene 99+%, Spectrophotometric Grade

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: TetrachloroethenePCEEthylene tetrachloridePerchloroethylene

167890025 167890010 167890100
TS167890010EA 191 USD
TS167890010 TS167890025 TS167890100
Tetrachloroethylene 99+%, Spectrophotometric Grade
Tetrachloroethylene
Formula: C₂Cl₄
MW: 165.83 g/mol
Boiling Pt: 120…122 °C (760 mmHg)
Melting Pt: –22 °C
Density: 1.625 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000834
CAS Number: 127-18-4
EINECS: 204-825-9
UN: 1897
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Water ≤0.05 %
GC ≥99.0 %
UV 1 cm cell vs water
UV at 290 nm A: ≤1
UV at 295 nm A: ≤0.3
UV at 305 nm A: ≤0.1
UV at 350 nm A: ≤0.05
UV at 400 nm A: ≤0.03
Color scale ≤10 APHA
Residue after evaporation ≤0.0005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR