Formic acid 98+%, pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Methanoic acidAminic Acid

147930100
TS147930100EA 266 USD
TS147930100
Formic acid 98+%, pure
Formic acid
Formula: HCO₂H
MW: 46.03 g/mol
Boiling Pt: 101 °C (760 mmHg)
Melting Pt: 8 °C
Density: 1.22 g/cm³
Flash Pt: 50 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003297
CAS Number: 64-18-6
EINECS: 200-579-1
UN: 1779
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3700 to 1.3720 (20°C, 589 nm)
Color scale ≤20 APHA
Titration with NaOH ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR