Petroleum spirit, 180…280 °C ∼, pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Gasoline, Petroleum ether, Petrol, Benzine

387490010 387490025 387490050 387490250
TS387490010EA 51 USD
TS387490010 TS387490025 TS387490050 TS387490250
Petroleum spirit, 180…280 °C ∼, pure
Petroleum spirit
Boiling Pt: 200…250 °C (760 mmHg)
Density: 0.79 g/cm³
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS Number: 64742-49-0
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4300 to 1.4500 (20°C, 589 nm)
Boiling point 200 to 250°C
Density (15°C) ≥0.79 kg/dm³
Aromatics ≤0.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR