Petroleum spirit, 60…95 °C for analysis
Supplier: Thermo Scientific Chemicals
|
Danger
|
235070010
235070025
TS235070010EA
76
USD
TS235070010
TS235070025
Petroleum spirit, 60…95 °C for analysis
Petroleum spirit
Boiling Pt:
60…95 °C (760 mmHg) Melting Pt: –40 °C Density: 0.7 g/cm³ Flash Pt: –20 °C Storage Temperature: Ambient |
MDL Number:
MFCD00081849 CAS Number: 64742-49-0 UN: 3295 ADR: 3,II |
Specification Test Results
Appearance | Clear colorless Liquid |
Water | ≤0.02 % (Coulometric) |
Boiling point | 60 to 95 °C (760 mmHg) |
Non-volatile matter | ≤0.001 % |
Acidity | ≤0.001 meq/g |
Copper (Cu) | ≤0.00002 % |
Iron (Fe) | ≤0.00005 % |
Lead (Pb) | ≤0.00002 % |
Nickel (Ni) | ≤0.00002 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...