Petroleum spirit, 35…60 °C ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Gasoline, Petroleum ether, Petrol, Benzine

422990050 422995000
TS422990050EA 218 USD
TS422990050 TS422995000
Petroleum spirit, 35…60 °C ACS
Petroleum spirit
Boiling Pt: 35…60 °C (1013 hPa)
Flash Pt: –30 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00081849
CAS Number: 64742-49-0
UN: 1268
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear liquid
Boiling point 35 to 60 °C
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %
Acidity Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR