Petroleum spirit, 40…60 °C, extra dry (max. 50 ppm H₂O), AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Gasoline, Petroleum ether, Petrol, Benzine

396920010 396921000
TS396920010EA 114 USD
TS396920010 TS396921000
Petroleum spirit, 40…60 °C, extra dry (max. 50 ppm H₂O), AcroSeal™
Petroleum spirit
Boiling Pt: 40…65 °C (760 mmHg)
Density: 0.65 g/cm³
Flash Pt: –45 °C
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS Number: 64742-49-0
UN: 1268
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.005 % (Coulometric)
Residue after evaporation ≤0.001 %
Filtered to 0.2 micrometer Filtered


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR