Petroleum spirit, 60…95 °C, extra_pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Gasoline, Petroleum ether, Petrol, Benzine
292510010
292510025
292510100
TS292510010EA
75
USD
TS292510010
TS292510025
TS292510100
Petroleum spirit, 60…95 °C, extra_pure
Petroleum spirit
Boiling Pt:
60…95 °C (760 mmHg) Melting Pt: –40 °C Density: 0.7 g/cm³ Flash Pt: –20 °C Storage Temperature: Ambient |
MDL Number:
MFCD00081849 CAS Number: 64742-49-0 EINECS: 265-151-9 UN: 3295 ADR: 3 II |
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3880 to 1.3920 (20°C, 589 nm) |
Boiling point | 60 to 95°C |
Density | (15°C) 0.690 to 0.710 g/ml |
Color scale | ≤10 APHA |
Residue after evaporation | ≤0.005 % |
n-Hexane | ≤2 % |
Benzene | ≤35 ppm |
Aromatics | ≤0.01 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...