Hydrochloric acid ~37% in water for biochemistry
Supplier: Thermo Scientific Chemicals
|
Danger
|
463961000
463965000
TS463961000EA
30
USD
TS463961000
TS463965000
Hydrochloric acid ~37% in water for biochemistry
Hydrochloric acid
- Acid
Formula:
HCl Boiling Pt: 57 °C (760 mmHg) Melting Pt: –35 °C Density: 1.18 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00011324 CAS Number: 7647-01-0 UN: 1789 ADR: 8,II |
Specification Test Results
Appearance | Clear solution |
Color scale | ≤10 APHA |
Residue after ignition | ≤0.0005 % |
Titration with NaOH | 36.5 to 38.0 % |
Bromide (Br) | ≤50 ppm |
Sulfite (SO3) | ≤1 ppm |
Sulfate (SO4) | ≤1 ppm |
Free chlorine | ≤1 ppm (as Cl) |
Ammonium (NH4) | ≤3 ppm |
Arsenic (As) | ≤0.01 ppm |
Iron (Fe) | ≤0.2 ppm |
Heavy metals (as Pb) | ≤1 ppm |
Extractable organic subst | ≤5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...