Hydrogen chloride 4->6 N in 2-propanol, technical

Supplier: Thermo Scientific Chemicals
Danger

450720010 450720025
TS450720010EA 89 USD
TS450720010 TS450720025
Hydrogen chloride 4->6 N in 2-propanol, technical
Hydrogen chloride
Formula: HCl
Density: 0.909 g/cm³
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 2920
ADR: 8,I

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Solution
Water ≤5 %
Strength 4 to 6 N


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR