Hydrogen chloride 1.25 M in ethanol, AcroSeal®
Supplier: Thermo Scientific Chemicals
|
Danger
|
427011000
427018000
TS427011000EA
59
USD
TS427011000
TS427018000
Hydrogen chloride 1.25 M in ethanol, AcroSeal®
Hydrogen chloride
Formula:
HCl Flash Pt: 12 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00011324 CAS Number: 7647-01-0 EINECS: 231-595-7 UN: 2924 ADR: 3,II |
Specification Test Results
Appearance | Clear colorless Solution |
Water | ≤0.5 % |
Strength | 1.10 to 1.40 M |
NMR | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...