n-Hexane, dry_extra 96+%, AcroSeal®

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Hexyl hydride, Hexanen, Hexane

326920010 326920025 326921000
TS326920010EA 120 USD
TS326920010 TS326920025 TS326921000
n-Hexane, dry_extra 96+%, AcroSeal®
n-Hexane
Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 69
Melting Pt: -94.3
Density: 0.659 g/cm³
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
Merck Index: 13,04712

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.002 %
GC ≥96.0 %
Residue after evaporation ≤0.0005 %
Filt.over 0.2 micron PFTE Filtered


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR