Diisopropylphosphine 10% (w/w) in hexanes

Supplier: Thermo Scientific Chemicals
Danger

383400025 383400100 383400500
TS383400025EA 93 USD
TS383400025 TS383400100 TS383400500
Diisopropylphosphine 10% (w/w) in hexanes
Diisopropylphosphine

Appearance: Clear colorless Solution

Formula: C₆H₁₅P
Density: 0.67 g/cm³
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD00233814
CAS Number: 20491-53-6
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Solution
NMR (31P-NMR) Consistent with structure
Concentration 9 to 11 wt%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR