Ammonium thioglycolate 70% (w/w) in water

Supplier: Thermo Scientific Chemicals
Danger

384680010 384682500
TS384680010EA 83 USD
TS384680010 TS384682500
Ammonium thioglycolate 70% (w/w) in water
Ammonium thioglycolate
Formula: C₂H₇NO₂S
Boiling Pt: 115 °C (760 mmHg)
Melting Pt: –20 °C
Density: 1.22 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00137451
CAS Number: 5421-46-5
UN: 2922
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless Solution
Titration Iodimetric 68.0 to 72.0 %
Identification mercaptane:positive
pH 5.8 to 6.4
Iron (Fe) ≤0.2 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR