Dess-Martin periodinane 15% (w/w) in dichloromethane

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,1-Dihydro-1,1,1-triacetoxy-1,2-benziodoxol-3(1H)-one, DMP, (1,1,1-Triacetoxy)-1,1-dihydro-1,2-benziodoxol-3(1H)-one, Acetic acid 1,1-diacetoxy-3-oxo-1l5-ioda-2-oxa-indan-1-yl ester

333110100 333110500 333112500
TS333110100EA 73 USD
TS333110100 TS333110500 TS333112500
Dess-Martin periodinane 15% (w/w) in dichloromethane
Dess-Martin periodinane

Appearance: Colorless to light yellow Solution

Formula: C₁₃H₁₃IO₈
Density: 1.362 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00130127
CAS Number: 87413-09-0
UN: 1593
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Colorless to light yellow Solution (May develop some turbidity or precipitate)
Titration Iodometric 10 to 15 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR