(±)-α-Terpineol 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-(4-Methylcyclohex-3-en-1-yl)propan-2-ol
, Terpineol
, α-Terpineol
301610025
301610250
301615000
TS301610025EA
197
USD
TS301610025
TS301610250
TS301615000
(±)-α-Terpineol 97+%
(±)-α-Terpineol
Formula:
C₁₀H₁₈O MW: 154.25 g/mol Boiling Pt: 214…224 °C (760 mmHg) Melting Pt: 31…34 °C Density: 0.937 g/cm³ Flash Pt: 90 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001557 CAS Number: 98-55-5 EINECS: 202-680-6 |
Specification Test Results
Appearance | Clear colorless to white solid |
Infrared spectrum | Conforms |
Refractive index | 1.4800 to 1.4850 (20°C, 589 nm) |
GC | ≥97.0 % |
Specific gravity | (20°C) 0.934 to 0.940 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...