Molecular sieve A5 (0.5 nm, 5 Å) 4-8 mesh
Supplier: Thermo Scientific Chemicals
Synonyms:
Linde 5A, Molecular sieves, 5A 3.0mm-5.0mm pellets
197280025
197285000
TS197280025EA
241
USD
TS197280025
TS197285000
Molecular sieve A5 (0.5 nm, 5 Å) 4-8 mesh
Molecular sieve A5 (0.5 nm, 5 Å)
MDL Number:
MFCD00131613 CAS Number: 69912-79-4 |
Specification Test Results
Appearance | Off-white to beige Beads |
Particle size | 4 to 8 mesh |
Bulk density | 660 to 780 g/l |
Residual water | ≤2 % (550°C) |
Water adsorption | ≥20 % (60% rH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...