Perchloric acid ∼70% in water for analysis
Supplier: Thermo Scientific Chemicals
|
Danger
|
223310010
223312500
TS223310010EA
307
USD
TS223310010
TS223312500
Perchloric acid ∼70% in water for analysis
Perchloric acid
Formula:
HClO₄ Boiling Pt: 203 °C (760 mmHg) Melting Pt: –18 °C Density: 1.66 g/cm³ Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011325 CAS Number: 7601-90-3 EINECS: 231-512-4 UN: 1873 ADR: 5.1,I |
Specification Test Results
Appearance | Clear liquid |
Color scale | ≤10 APHA |
Ash | ≤0.003 % |
Titration with NaOH | 70 to 72 % |
Chloride (Cl) | ≤10 ppm |
Sulfate (SO4) | ≤10 ppm |
Silicate (as SiO2) | ≤0.0005 % |
Iron (Fe) | ≤1 ppm |
Heavy metals (as Pb) | ≤1 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...