Benzyl alcohol, meets analytical specification of Ph.Eur.

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Benzenemethanol, Phenylmethanol, Phenylcarbinol

447000010 447000025
TS447000010EA 121 USD
TS447000010 TS447000025
Benzyl alcohol, meets analytical specification of Ph.Eur.
Benzyl alcohol
Formula: C₆H₅CH₂OH
MW: 108.14 g/mol
Boiling Pt: 205
Melting Pt: -15.3
Density: 1.044 g/cm³
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00004599
CAS Number: 100-51-6
Merck Index: 13,01126

Order Now

Specification Test Results

Appearance Clear colorless Oily liquid
Infrared spectrum Conforms
Refractive index 1.5380 to 1.5410 (20°C, 589 nm)
Assay 98.0 to 100.5 %
Appearance of solution Clear colorless
Residue after evaporation ≤0.05 %
Related substances ≤0.3 % (Sum of other peaks with relative retention greater than that of benzylalcohol)
Acidity Passes test
Peroxide value ≤5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR