Benzyl alcohol, extra dry 98+%, AcroSeal®

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Benzenemethanol, Phenylmethanol, Phenylcarbinol

396880010 396881000
TS396880010EA 260 USD
TS396880010 TS396881000
Benzyl alcohol, extra dry 98+%, AcroSeal®
Benzyl alcohol
Formula: C₆H₅CH₂OH
MW: 108.14 g/mol
Boiling Pt: 205
Melting Pt: -15.3
Density: 1.044 g/cm³
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00004599
CAS Number: 100-51-6
Merck Index: 13,01126

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥98.0 %
Residue after evaporation ≤0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR