Potassium-tert-butoxide 1 M in tert-butanol, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Potassium tert-butanolatePotassium t-butoxidePotassium tert-butoxidePotassium tert-butylate

426128000
TS426128000EA 396 USD
TS426128000
Potassium-tert-butoxide 1 M in tert-butanol, AcroSeal™
Potassium-tert-butoxide
Formula: C₄H₉KO
MW: 112.21 g/mol
Density: 0.82 g/cm³
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00012162
CAS Number: 865-47-4
UN: 2920
ADR: 8,I

Order Now

Specification Test Results

Appearance Colorless to amber Solution
Strength 0.95 to 1.10 M
Refractive index 1.5750 to 1.5780 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR