Sulfuryl chloride 1.0 M in dichloromethane, AcroSeal®
Supplier: Thermo Scientific Chemicals
|
Danger
|
378151000
378158000
TS378151000EA
66
USD
TS378151000
TS378158000
Sulfuryl chloride 1.0 M in dichloromethane, AcroSeal®
Sulphuryl chloride
Formula:
Cl₂O₂S Storage Temperature: Ambient |
MDL Number:
MFCD00011451 CAS Number: 7791-25-5 EINECS: 232-245-6 UN: 2922 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless to yellow Solution |
Titration Argentometric | 0.95 to 1.10 M |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...