Potassium hydroxide 1 N ethanol

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Potash caustic, Potassium hydrate, Caustic potash

393320010 393322500
TS393320010EA 48 USD
TS393320010 TS393322500
Potassium hydroxide 1 N ethanol
Potassium hydroxide
Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1320
Melting Pt: 360
Density: 0.86 g/cm³
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
UN: 2924
ADR: 3 II
Merck Index: 12,07806

Order Now

Specification Test Results

Appearance Clear colorless to yellow Solution (May darken on storage)
Strength 0.950 to 1.050 N


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR