Sodium hydroxide solution_standard 0.2 mol/l (0.2 N) water

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

349680025 349685000
TS349680025EA 62 USD
TS349680025 TS349685000
Sodium hydroxide solution_standard 0.2 mol/l (0.2 N) water
Sodium hydroxide
Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390
Melting Pt: 323
Density: 1.01 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
Merck Index: 14,08627

Order Now

Specification Test Results

Appearance Clear colorless Solution
Strength 0.1996 to 0.2004 N


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR