Water ACS

Supplier: Thermo Scientific Chemicals

Synonyms: Dihydrogen oxide

434400010 434400050
TS434400010EA 51 USD
TS434400010 TS434400050
Water ACS
Water
Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
Density: 1 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Merck Index: 13,10098

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Chloride (Cl) ≤0.4 ppm
Nitrate (NO3) ≤0.4 ppm
Phosphate (PO4) ≤1.0 ppm
Sulfate (SO4) ≤1.0 ppm
Silicate (as SiO2) ≤0.01 ppm
Heavy metals (as Pb) ≤0.01 ppm
KMnO4-reducing substance Passes test
Conductivity 2.0 x 10-6 ohm-1 cm-1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR