Sodium chloride ∼26% water, pure
Supplier: Thermo Scientific Chemicals
Synonyms:
Saline, NaCl, Sea salt, Halite
387640010
387640025
TS387640010EA
45
USD
TS387640010
TS387640025
Sodium chloride ∼26% water, pure
Sodium chloride
Formula:
NaCl MW: 58.44 g/mol Boiling Pt: 1413 Melting Pt: 801 Density: 1.202 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00003477 CAS Number: 7647-14-5 Merck Index: 14,08599 |
Specification Test Results
Appearance | Clear colorless Solution |
Titration Argentometric | 25 to 28 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...