Sodium borohydride 12% in sodium hydroxide solution 40%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sodium tetrahydridoborate, Sodium tetrahydroborate

389930010 389930025 389931000 389932500
TS389930010EA 98 USD
TS389930010 TS389930025 TS389931000 TS389932500
Sodium borohydride 12% in sodium hydroxide solution 40%
Sodium borohydride
Formula: NaBH₄
Storage Temperature: Ambient
MDL Number: MFCD00003518
CAS Number: 16940-66-2
UN: 3320
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless Liquid
Titration Iodometric 11 to 13 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR