Methylcyclohexane, extra dry 98+%, AcroSeal™
Supplier: Thermo Scientific Chemicals
|
Danger
|
397240010
397241000
TS397240010EA
213
USD
TS397240010
TS397241000
Methylcyclohexane, extra dry 98+%, AcroSeal™
Methylcyclohexane
Formula:
C₆H₁₁CH₃ Boiling Pt: 101 °C (760 mmHg) Melting Pt: –126.3 °C Density: 0.77 g/cm³ Flash Pt: –3 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001497 CAS Number: 108-87-2 UN: 2296 ADR: 3,II |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Water | ≤0.005 % |
GC | ≥98.0 % |
Residue after evaporation | ≤0.001 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...