Cyclopentane 95+%, pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Pentamethylene

397430010 397430050
TS397430010EA 171 USD
TS397430010 TS397430050
Cyclopentane 95+%, pure
Cyclopentane
Formula: C₅H₁₀
MW: 70.13 g/mol
Boiling Pt: 50 °C (760 mmHg)
Melting Pt: –94 °C
Density: 0.75 g/cm³
Flash Pt: –7 °C
Storage Temperature: Ambient
MDL Number: MFCD00001356
CAS Number: 287-92-3
UN: 1146
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR