Cyclohexane, extra dry over Molecular Sieve 99.5%, AcroSeal®
Supplier: Thermo Scientific Chemicals
|
Danger
|
364660010
364660025
364661000
364665000
TS364660010EA
108
USD
TS364660010
TS364660025
TS364661000
TS364665000
Cyclohexane, extra dry over Molecular Sieve 99.5%, AcroSeal®
Cyclohexane
Formula:
C₆H₁₂ Boiling Pt: 81 °C (760 mmHg) Melting Pt: 6.5 °C Density: 0.77 g/cm³ Flash Pt: –18 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003814 CAS Number: 110-82-7 EINECS: 203-806-2 UN: 1145 ADR: 3,II |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Water | ≤0.005 % (Coulometric) |
GC | ≥99.4 % |
Acidity | ≤0.001 meq/g |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...