Methanol 99.8%, ECD tested for pesticide analysis for residue analysis

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Wood alcohol, Methyl alcohol, Methyl hydroxide, Hydroxy methane, Carbinol, Methylol

326630010 326630025
TS326630010EA 55 USD
TS326630010 TS326630025
Methanol 99.8%, ECD tested for pesticide analysis for residue analysis
Methanol
Formula: CH₃OH
MW: 32.04 g/mol
Boiling Pt: 64.6
Melting Pt: -98
Density: 0.791 g/cm³
Flash Pt: 12 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3 II
REACH: 01-2119433307-44
Merck Index: 13,05984

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.1 % (Coulometric)
GC ≥99.8 %
Color scale ≤10 APHA
GC/ECD (as lindane) ≤5 ng/l
Residue after evaporation ≤0.0002 %
Acidity (CH3COOH) ≤0.003 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR