Acetic acid 99+%, extra pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Vinegar acid, Methanecarboxylic acid, Ethanoic acid
295310050
295310250
TS295310050EA
198
USD
TS295310050
TS295310250
Acetic acid 99+%, extra pure
Acetic acid
Formula:
H₃CCOOH MW: 60.05 g/mol Boiling Pt: 118 Melting Pt: 17 Density: 1.048 g/cm³ Flash Pt: 40 °C Storage Temperature: Ambient |
MDL Number:
MFCD00036152 CAS Number: 64-19-7 UN: 2789 ADR: 8 II |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3700 to 1.3730 (20°C, 589 nm) |
Water | ≤0.04 % |
Residue after evaporation | ≤0.01 % |
Titration with NaOH | ≥99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...