Chicken Embryology (Gallus domesticus) - English Slides
Supplier: AMERICAN 3B SCIENTIFIC SE
1003986
470357-036EA 375 USD
470357-036
Chicken Embryology (Gallus domesticus) - English Slides
Slides Prepared Slides
Ten microscope slides.
1(f). Chicken, 24 hour, t.s. with neural groove, notochord, germ layers
2(f). Chicken, 36 hour, t.s. with neural tube
3(g). Chicken, 48 hour, l.s. with differentiation of mesoderm and ectoderm
4(f). Chicken, 3 days, t.s. through body showing amnion and serosa
5(f). Chicken, 3 days, t.s. of head with primordium of brain, eyes and heart
6(g). Chicken, 3 to 4 days, horizontal section of entire specimen shows primordia of organs
7(f). Chicken, 4 to 5 days, t.s. region of head with brain, gill arches
8(f). Chicken, 4 to 5 days, t.s. region of heart
9(g). Chicken, 8 days, sagittal l.s. through entire specimen showing embryonic organs
10(f). Chicken, feather development, sec. through wings