Crystal Digital PCR® Assay for BRAF/KRAS/NRAS
Supplier: STILLA TECHNOLOGIES, INC.
R51006.1 R51006.2
77590-078EA 575.36 USD
77590-078 77590-080
Crystal Digital PCR® Assay for BRAF/KRAS/NRAS
Assays qPCR Assays
Assay-specific crystal flex probes and primers for the detection of BRAF/KRAS/NRAS mutations and its positive control. The BRAF/KRAS/NRAS Crystal Digital PCR® assay is a 10X assay designed to detect and quantify mutations in BRAF, KRAS, and NRAS genes using the ruby chip. BRAF, KRAS, and NRAS genes are pivotal in regulating cell signaling pathways implicated in cancer development.
  • Ready-to-use assay reagents to detect multiple targets in a single reaction
  • Crystal flex probe detection chemistry for flexible assay configuration
  • Same set of crystal universal reporters to detect any target in different assays
Wet lab tested
Compatibility: Nio™ Digital PCR, Prism6, ruby chip
Channels: Blue, teal, green, yellow, red, infra-red
Required reagents: Crystal universal reporter 7 and naica® PCR MIX 10X
Targets: BRAF V600-K601, KRAS G12-G13, NRAS G12-G13
Caution: For research use only. Not for use in diagnostic procedures.