Crystal Universal Reporters
Supplier: STILLA TECHNOLOGIES, INC.
R41401 R41402 R42401 R42402
77590-046EA 753.45 USD
77590-046 77590-048 77590-050 77590-052
Crystal Universal Reporters
Assays qPCR Assays
Crystal universal reporters generate fluorescent signals when activated during a Crystal Digital PCR® reaction using target-specific crystal flex probe detection chemistry.
  • Select the crystal universal reporter compatible with the Crystal Digital PCR® Assay of interest, either in a 200 or a 1000 ruby chip reaction format
Their universal design allows them to be used with any Crystal Digital PCR® assay, providing a modular approach to detection. This means you can use the same set of crystal Universal Reporters to detect and quantify all targets of interest, regardless of the specific assay or the degree of multiplexing.