Solvent Absorption Concentrators
Supplier: Sartorius
Vivapore®
VP-S010P0008-3 VP-S005P0008-3 VP-S010P0008-3 VP-S005P0008-3
77581-126PK 226.53 USD
77581-126 CA77581-128 77581-128 CA77581-126
Solvent Absorption Concentrators
Concentrators
Vivapore solvent absorption concentrators are disposable ultrafiltration units for the concentration of macromolecules from aqueous samples.
  • Simply fill and go, for equipment-free protein concentration
  • Gentle processing of shear-sensitive samples
  • Designed to achieve high concentration factors
Concentrate equipment-free solvent absorption is an economic and user-friendly technique for sample concentration in research and clinical laboratories, since there is no need for additional equipment such as centrifuges or pressure regulators.
Vivapore® 10 can be used with an expansion reservoir to increase the sample capacity from 10 to 20 ml. Alternatively, the expansion reservoir may be used to desalt samples up to 2 ml in volume. When filled with an appropriate exchange buffer, this will replace the original solvent at the same rate as ultrafiltration.
Device type: Solvent absorption
Housing material: Styrene acrylonitrile (SAN)
Delivery information: Each pack of Vivapore® includes one stand to hold four units during use. In addition, each pack of Vivapore® ten includes one expansion reservoir. If you require additional stands and reservoirs, these are available to purchase separately (order numbers 77581-124 and 77581-122).