Buccutite™ Rapid PE-Texas Red Tandem Antibody Labelling Kit
Supplier: AAT Bioquest
Buccutite™
5412
77462-884EA 1811.25 USD
77462-884
Buccutite™ Rapid PE-Texas Red Tandem Antibody Labelling Kit
Protein Linking and Labeling Systems
Production scale optimized for labelling 1 mg antibody per reaction. Buccutite™ Rapid PE-Texas Red tandem antibody labelling kits, designed for large-scale production, provide a streamlined approach for labelling antibodies with PE, APC, PerCP and iFluor® tandem dyes.
  • Robust protocol
  • Consistent results
  • Six-month shelf life
Designed to provide high quality material for downstream fluorescent applications, including flow cytometry.
Delivery information: Shipped with blue ice.
Caution: Research use only.