Ytterbium(III) isopropoxide ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

Synonyms: Ytterbium(III) i-propoxide

Y0010-1G Y0010-200MG
TCY0010-1GEA 290.46 USD
TCY0010-1G TCY0010-200MG
Ytterbium(III) isopropoxide ≥98.0% (by titrimetric analysis)
Ytterbium (III) isopropoxide
Formula: C₉H₂₁O₃Yb
MDL Number: MFCD00145532
CAS Number: 6742-69-4
UN: 2925
ADR: 4.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e72408616bc336c17a43241920f02793104fadf3&vid=1df27f6aa783a310a2e9b7f8b5b1c5ae53aa7821&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR