2-(p-Tolyl)pyrimidine ≥98.0% (by GC)

Supplier: TCI America
Warning

T3759-1G T3759-200MG
TCT3759-1GEA 236.9 USD
TCT3759-1G TCT3759-200MG
2-(p-Tolyl)pyrimidine ≥98.0% (by GC)
2-(p-Tolyl)pyrimidine
Formula: C₁₁H₁₀N₂
MDL Number: MFCD08701658
CAS Number: 77232-13-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd21fefc0266082250a605bb8715f98f457a3f3b&vid=1d7e5e8e194998df9a5666c42984c30b58c9ea0c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 88.0 to 92.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR