4,4,5,5-Tetramethyl-2-(10-phenylanthracen-9-yl)-1,3,2-dioxaborolane ≥98.0% (by GC)

Supplier: TCI America

T3665-1G T3665-5G
TCT3665-1GEA 44.29 USD
TCT3665-1G TCT3665-5G
4,4,5,5-Tetramethyl-2-(10-phenylanthracen-9-yl)-1,3,2-dioxaborolane ≥98.0% (by GC)
4,4,5,5-Tetramethyl-2-(10-phenylanthracen-9-yl)-1,3,2-dioxaborolane
Formula: C₂₆H₂₅BO₂
MDL Number: MFCD13194955
CAS Number: 460347-59-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ed3e75691567d34bfa7a475c73b32209176ccfbb&vid=ee2d0d292b8b608edb95d2f6a16c8b88e0251750&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 165.0 to 169.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR