Polyethylene glycol monododecyl ether n=∼23

Supplier: TCI America
Warning

Synonyms: Polidocanol

P2737-25G P2737-500G
TCP2737-25GEA 29.87 USD
TCP2737-25G TCP2737-500G
Polyethylene glycol monododecyl ether n=∼23
Polyethylene glycol monododecyl ether
Formula: HO(CH₂CH₂O)nC₁₂H₂₅
MDL Number: MFCD00080891
CAS Number: 9002-92-0
EINECS: 500-002-6
UN: 3077
ADR: 9,III
Merck Index: 14,07558

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Freezing point 33.0 to 38.0 deg-C
The average value of n 21.0 to 25.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR