SMER28 ≥98.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: N-Allyl-6-bromoquinazolin-4-amine

M3106-100MG M3106-25MG
TCM3106-100MGEA 557.23 USD
TCM3106-100MG TCM3106-25MG
SMER28 ≥98.0% (by HPLC)
SMER28
Formula: C₁₁H₁₀BrN₃
Storage Temperature: Refrigerator
MDL Number: MFCD02166825
CAS Number: 307538-42-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=adeea31f6985f713f680b5b09e3ff26116e748ad&vid=1ab4979858bd340a22b8eebe4083908a733cf950&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 15.00 to 16.70 %
Melting point 167.0 to 171.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR