4-Phenylpyrimidine ≥96.0% (by GC, titration analysis)

Supplier: TCI America
Warning

P2525-1G P2525-5G
TCP2525-1GEA 55.62 USD
TCP2525-1G TCP2525-5G
4-Phenylpyrimidine ≥96.0% (by GC, titration analysis)
4-Phenylpyrimidine
Formula: C₁₀H₈N₂
Boiling Pt: 144 °C
MDL Number: MFCD00006111
CAS Number: 3438-48-0
EINECS: 222-345-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea8e2d59ac4114e86dece0f45bb1a2b54ef8ff1c&vid=abe09a54f734181d85572737c54567b31a49e396&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 96.0 %
Purity (Nonaqueous Titration) min. 96.0 %
Melting point 60.0 to 64.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR