Monarch® Spin Plasmid Miniprep Kits
Supplier: New England Biolabs (NEB)
T1110L T1110S
77575-356EA 392.77 USD
77575-356 77575-358
Monarch® Spin Plasmid Miniprep Kits
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The Monarch® Spin Plasmid Miniprep kit is a newer version of the original Monarch® Miniprep kit, and offers a unique method for plasmid preparation and purification. This kit enables extraction of highly concentrated plasmid DNA from various bacterial strains, with yields of up to 20 µg of high-quality DNA.
  • High purity in the purification of plasmid DNA
  • Elute in as low as 30 µl, allowing for highly concentrated plasmid DNA
  • Unique column design enables elution in low volumes and minimizes contaminant carry-over
  • Optimized buffer system with color indicators to monitor the completion of each step
  • Purified plasmid DNA is ready for restriction digestion and other downstream applications
This kit features upgraded spin columns, precision-engineered to uniquely allow for low elution (as little as 30 µl), for highly concentrated DNA in downstream application such as sequencing, restriction digestion, transformation, transfection, and more. A colored buffer system helps indicate completion of key lysis and neutralization steps for an easy to follow workflow and shorter, faster protocols allow for reduced hands-on time and less spin time. Compared to leading suppliers, these kits use significantly less plastic through uniquely designed columns and bottles with plastic reduction while still maintaining structural integrity for centrifugation or vacuum manifolds and other steps.